• USA Home
  • Search Results

  • methylaminesolution31067489511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methylamine solution

4 Products
Synonym:   Monomethylamine
Image of Methylamine solution
CAS Number:
Linear Formula:
 CH3NH2
Molecular Weight:
 31.06
 Beilstein/REAXYS Number:
 741851
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Vapor pressure
  Refractive Index
  Greener Alternative Principles
  Density
  Concentration
Show More