• USA Home
  • Search Results

  • methylbdxylopyranoside1641661205511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl β-D-xylopyranoside

1 Products
Image of Methyl β-<SC>D</SC>-xylopyranoside
CAS Number:
Empirical Formula (Hill Notation):
 C6H12O5
Molecular Weight:
 164.16
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More