• USA Home
  • Search Results

  • methylbromoacetate152979632211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl bromoacetate

2 Products
Image of Methyl bromoacetate
CAS Number:
Linear Formula:
 BrCH2COOCH3
Molecular Weight:
 152.97
 Beilstein/REAXYS Number:
 506256
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More