• USA Home
  • Search Results

  • methylbutyrate1021362342711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl butyrate

5 Products
Image of Methyl butyrate
CAS Number:
Linear Formula:
 CH3CH2CH2COOCH3
Molecular Weight:
 102.13
 Beilstein/REAXYS Number:
 1740743
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Agency/Method
  Document Links
  Shelf Life
  Allergen
  Organoleptic property
  Special Grade
  Application(s)
  Solubility
  Assay
Show More