• USA Home
  • Search Results

  • methylcarbamate750759855011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl carbamate

1 Products
Synonym:   Urethylane
Image of Methyl carbamate
CAS Number:
Linear Formula:
 NH2COOCH3
Molecular Weight:
 75.07
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Solubility
Show More

Product #
Description
246352 Methyl carbamate, 98%,