• USA Home
  • Search Results

  • methylcyclohexane981910887211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methylcyclohexane

5 Products
Synonym:   Hexahydrotoluene
Image of Methylcyclohexane
CAS Number:
Linear Formula:
 C6H11CH3
Molecular Weight:
 98.19
 Beilstein/REAXYS Number:
 505972
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Refractive Index
  Application(s)
  Purity Grade
  Special Grade
  UV-Absorption Values
Show More