• USA Home
  • Search Results

  • methyldihydrojasmonatemixtureofcisandtrans226312485198711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl dihydrojasmonate, mixture of cis and trans

2 Products
Synonym:   (3-Oxo-2-pentylcyclopentyl)acetic acid methyl ester, Methyl (3-oxo-2-pentylcyclopentyl)acetate
Image of Methyl dihydrojasmonate, mixture of <I>cis </I>and <I>trans</I>
CAS Number:
Empirical Formula (Hill Notation):
 C13H22O3
Molecular Weight:
 226.31
 Council of Europe no.:
 10785
 EC Number:
 FEMA Number:
 3408
 Flavis number:
 9.520
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Assay
  Solubility
Show More