• USA Home
  • Search Results

  • methylelaidate29649193762811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl elaidate

2 Products
Synonym:   Elaidic acid methyl ester, Methyl trans-9-octadecenoate
Image of Methyl elaidate
CAS Number:
Linear Formula:
 CH3(CH2)7CH=CH(CH2)7COOCH3
Molecular Weight:
 296.49
 Beilstein/REAXYS Number:
 1727038
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Functional group
  Physical Form
  Application(s)
  Ice Shipping
Show More