• USA Home
  • Search Results

  • methylenebluesolution319856173411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methylene Blue solution

3 Products
Synonym:   Ehrlich’s reagent III, Loeffler’s Methylene Blue, Methylene blue stain
Image of Methylene Blue solution
CAS Number:
Empirical Formula (Hill Notation):
 C16H18ClN3S
Molecular Weight:
 319.85
 Beilstein/REAXYS Number:
 3641570
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Shelf Life
  Physical Form
  Density
  Special Grade
  Application(s)
Show More