• USA Home
  • Search Results

  • methylethanesulfonate12416191228311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl ethanesulfonate

1 Products
Synonym:   Ethanesulfonic acid methyl ester
Image of Methyl ethanesulfonate
CAS Number:
Linear Formula:
 C2H5SO3CH3
Molecular Weight:
 124.16
 Beilstein/REAXYS Number:
 1747452
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More