• USA Home
  • Search Results

  • methylethylketone72117893311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl ethyl ketone

3 Products
Synonym:   2-Butanone, Ethyl methyl ketone, MEK, Methyl ethyl ketone
Image of Methyl ethyl ketone
CAS Number:
Linear Formula:
 C2H5COCH3
Molecular Weight:
 72.11
 Beilstein/REAXYS Number:
 741880
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
Show More