• USA Home
  • Search Results

  • methylglyoxalsolution72067898811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methylglyoxal solution

2 Products
Synonym:   Acetylformaldehyde, Pyruvaldehyde, Pyruvic aldehyde
Image of Methylglyoxal solution
CAS Number:
Linear Formula:
 CH3COCHO
Molecular Weight:
 72.06
 Beilstein/REAXYS Number:
 906750
 MDL number:
 PubChem Substance ID:
Popular Documents:  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Density
Show More