• USA Home
  • Search Results

  • methylheptanoate1442110673011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl heptanoate

3 Products
Synonym:   Methyl enanthate, Heptanoic acid methyl ester
Image of Methyl heptanoate
CAS Number:
Empirical Formula (Hill Notation):
 C8H16O2
Molecular Weight:
 144.21
 EC Index Number
 203-428-8
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Titertest
  Assay
Show More