• USA Home
  • Search Results

  • methylhexanoate1301810670711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl hexanoate

6 Products
Synonym:   Methyl caproate, Caproic acid methyl ester, Hexanoic acid methyl ester
Image of Methyl hexanoate
CAS Number:
Linear Formula:
 CH3(CH2)4COOCH3
Molecular Weight:
 130.18
 EC Index Number
 203-425-1
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Anal. Standard Form
  Shelf Life
  Allergen
  Organoleptic property
  Physical Form
  Special Grade
  Titertest
  Assay
Show More