• USA Home
  • Search Results

  • methylisobutylketone1001610810111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl isobutyl ketone

3 Products
Synonym:   4-Methyl-2-pentanone, Isobutyl methyl ketone, Isopropylacetone, MIBK, Methyl isobutyl ketone
Image of Methyl isobutyl ketone
CAS Number:
Linear Formula:
 (CH3)2CHCH2COCH3
Molecular Weight:
 100.16
 Beilstein/REAXYS Number:
 605399
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
  Package quantity
Show More