• USA Home
  • Search Results

  • methyllaurate2143411182011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl laurate

7 Products
Synonym:   Lauric acid methyl ester, Methyl dodecanoate
Image of Methyl laurate
CAS Number:
Linear Formula:
 CH3(CH2)10CO2CH3
Molecular Weight:
 214.34
 Beilstein/REAXYS Number:
 1767780
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Refractive Index
  Document Links
  Organoleptic property
  Density
  Biological Source
  Allergen
  Agency/Method
  Assay
Show More