• USA Home
  • Search Results

  • methylmethanesulfonate110136627311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl methanesulfonate

3 Products
Synonym:   Methanesulfonic acid methyl ester
Image of Methyl methanesulfonate
CAS Number:
Linear Formula:
 CH3SO3CH3
Molecular Weight:
 110.13
 Beilstein/REAXYS Number:
 1098586
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More