• USA Home
  • Search Results

  • methyloleate2964911262911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl oleate

6 Products
Synonym:   Methyl cis-9-octadecenoate, Oleic acid methyl ester
Image of Methyl oleate
CAS Number:
Linear Formula:
 CH3(CH2)7CH=CH(CH2)7CO2CH3
Molecular Weight:
 296.49
 Beilstein/REAXYS Number:
 1727037
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Application(s)
  Potency
  Assay
Show More