• USA Home
  • Search Results

  • methylophiopogonanonea342347480592811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methylophiopogonanone A

1 Products
Synonym:   (3R)-3-(1,3-Benzodioxol-5-ylmethyl)-2,3-dihydro-5,7-dihydroxy-6,8-dimethyl-4H-1-benzopyran-4-one
Image of Methylophiopogonanone A
CAS Number:
Empirical Formula (Hill Notation):
 C19H18O6
Molecular Weight:
 342.34
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More