• USA Home
  • Search Results

  • methylophiopogonanoneb328367480591711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methylophiopogonanone B

2 Products
Synonym:   (3R)-2,3-Dihydro-5,7-dihydroxy-3-[(4-methoxyphenyl)methyl]-6,8-dimethyl-4H-1-benzopyran-4-one
Image of Methylophiopogonanone B
CAS Number:
Empirical Formula (Hill Notation):
 C19H20O5
Molecular Weight:
 328.36
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More