• USA Home
  • Search Results

  • methylpalmitate2704511239011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl palmitate

6 Products
Synonym:   n-Hexadecanoic acid methyl ester, Methyl hexadecanoate, Palmitic acid methyl ester
Image of Methyl palmitate
CAS Number:
Linear Formula:
 CH3(CH2)14CO2CH3
Molecular Weight:
 270.45
 Beilstein/REAXYS Number:
 1780973
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Shelf Life
  Functional group
  Physical Form
  Ice Shipping
  Biological Source
  Special Grade
  Application(s)
Show More