• USA Home
  • Search Results

  • methylpanisate1661712198211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl p-anisate

2 Products
Synonym:   p-Anisic acid methyl ester, Methyl 4-methoxybenzoate
Image of Methyl <I>p</I>-anisate
CAS Number:
Linear Formula:
 CH3OC6H4CO2CH3
Molecular Weight:
 166.17
 Beilstein/REAXYS Number:
 2208571
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Agency/Method
  Special Grade
  Assay
Show More