• USA Home
  • Search Results

  • methylparaben152159976311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl Paraben

4 Products
Synonym:   p-Hydroxybenzoic acid methyl ester, Methyl 4-hydroxybenzoate, Methyl paraben, NIPAGIN
Image of Methyl Paraben
CAS Number:
Linear Formula:
 HOC6H4CO2CH3
Molecular Weight:
 152.15
 Beilstein/REAXYS Number:
 509801
 E Number:
 E218
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Anal. Standard Form
  Physical Form
  Special Grade
Show More