• USA Home
  • Search Results

  • methylpentadecanoate25642713264111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl pentadecanoate

2 Products
Synonym:   Pentadecanoic acid methyl ester
Image of Methyl pentadecanoate
CAS Number:
Linear Formula:
 CH3(CH2)13CO2CH3
Molecular Weight:
 256.42
 Beilstein/REAXYS Number:
 1776383
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Functional group
  Physical Form
  Application(s)
  Ice Shipping
Show More