• USA Home
  • Search Results

  • methylphenidatehydrochloride2697729859911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methylphenidate hydrochloride

1 Products
Synonym:   threo-Methyl α-phenyl-α-(2-piperidyl)acetate hydrochloride, Ritalin hydrochloride
Image of Methylphenidate hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C14H19NO2 · HCl
Molecular Weight:
 269.77
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More