• USA Home
  • Search Results

  • methylprednisolone374478343211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methylprednisolone

4 Products
Synonym:   11β,17α,21-Trihydroxy-6α-methyl-1,4-pregnadiene-3,20-dione, 6α-Methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione, 6α-Methylprednisolone, Medrol, Medrone
Image of Methylprednisolone
CAS Number:
Empirical Formula (Hill Notation):
 C22H30O5
Molecular Weight:
 374.47
 Beilstein/REAXYS Number:
 2340300
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Physical Form
  Application(s)
  Anal. Standard Form
  Manufacturer designation
Show More