• USA Home
  • Search Results

  • methylpropargylether700962741811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl propargyl ether

1 Products
Synonym:   3-Methoxy-1-propyne, Methyl 2-propynyl ether
Image of Methyl propargyl ether
CAS Number:
Linear Formula:
 HC≡CCH2OCH3
Molecular Weight:
 70.09
 Beilstein/REAXYS Number:
 878166
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More