• USA Home
  • Search Results

  • methylredsolution2693049352711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl Red solution

1 Products
Image of Methyl Red solution
CAS Number:
Empirical Formula (Hill Notation):
 C15H15N3O2
Molecular Weight:
 269.30
 Beilstein Registry Number:
 1843037
 MDL number:
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Chemical Composition
Show More