• USA Home
  • Search Results

  • methylsalicylate1521511936811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl salicylate

10 Products
Synonym:   2-Hydroxybenzoic acid methyl ester, Methyl 2-hydroxybenzoate, Oil of wintergreen, Wintergreen oil
Image of Methyl salicylate
CAS Number:
Linear Formula:
 2-(HO)C6H4CO2CH3
Molecular Weight:
 152.15
 Beilstein/REAXYS Number:
 971516
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Document Links
  Allergen
  Organoleptic property
  Biological Source
  Anal. Standard Form
  Manufacturer designation
  Application(s)
  Product Line
  Agency/Method
  Assay
  Physical Form
Show More