• USA Home
  • Search Results

  • methylstearate2985011261811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl stearate

8 Products
Synonym:   Methyl octadecanoate, Stearic acid methyl ester
Image of Methyl stearate
CAS Number:
Linear Formula:
 CH3(CH2)16CO2CH3
Molecular Weight:
 298.50
 Beilstein/REAXYS Number:
 1786213
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Shelf Life
  Functional group
  Ice Shipping
  Assay
  Anal. Standard Form
  Physical Form
  Application(s)
Show More