• USA Home
  • Search Results

  • methyltestosterone302455818411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyltestosterone

2 Products
Synonym:   17α-Methyl-4-androsten-17β-ol-3-one, 17α-Methyltestosterone, 17β-Hydroxy-17α-methyl-4-androsten-3-one, Mesterone, Methyltestosterone
Image of Methyltestosterone
CAS Number:
Empirical Formula (Hill Notation):
 C20H30O2
Molecular Weight:
 302.45
 Beilstein/REAXYS Number:
 2057425
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Special Grade
Show More