• USA Home
  • Search Results

  • methylviolet12345800487311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Methyl violet

1 Products
Synonym:   Basic Violet 1, Methyl violet 2B
Image of Methyl violet
CAS Number:
 Beilstein/REAXYS Number:
 8464808
 Colour Index Number:
 42535
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More