• USA Home
  • Search Results

  • metribuzin214292108764911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Metribuzin

2 Products
Image of Metribuzin
CAS Number:
Empirical Formula (Hill Notation):
 C8H14N4OS
Molecular Weight:
 214.29
 Beilstein/REAXYS Number:
 746650
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Anal. Standard Form
  Shelf Life
  Physical Form
  Special Grade
Show More