• USA Home
  • Search Results

  • metronidazole1711544348111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Metronidazole

8 Products
Synonym:   2-Methyl-5-nitroimidazole-1-ethanol
Image of Metronidazole
CAS Number:
Empirical Formula (Hill Notation):
 C6H9N3O3
Molecular Weight:
 171.15
 Beilstein/REAXYS Number:
 611683
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Shelf Life
  Product Line
  Manufacturer designation
  Mode of Action
  Anal. Standard Form
  Physical Form
  Application(s)
  Activity Spectrum
Show More