• USA Home
  • Search Results

  • mettlertoledocalibrationsubstanceme18555benzoicacid122126585011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Mettler-Toledo Calibration substance ME 18555, Benzoic acid

1 Products
Synonym:   Benzoic acid
Image of Mettler-Toledo Calibration substance ME 18555, Benzoic acid
CAS Number:
Linear Formula:
 C6H5COOH
Molecular Weight:
 122.12
 Beilstein/REAXYS Number:
 636131
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More