• USA Home
  • Search Results

  • mettlertoledocalibrationsubstanceme18872caffeine194195808211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Mettler-Toledo Calibration substance ME 18872, Caffeine

1 Products
Synonym:   1,3,7-Trimethylxanthine, Caffeine
Image of Mettler-Toledo Calibration substance ME 18872, Caffeine
CAS Number:
Empirical Formula (Hill Notation):
 C8H10N4O2
Molecular Weight:
 194.19
 Beilstein/REAXYS Number:
 17705
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More