• USA Home
  • Search Results

  • mettlertoledocalibrationsubstanceme51143091saccharin183188107211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Mettler-Toledo Calibration substance ME 51143091, Saccharin

1 Products
Synonym:   2,3-Dihydroxy-1,2-benzisothiazol-3-one-1,1-dioxide, 2-Sulfobenzoic acid imide, o-Benzoic sulfimide, Saccharin
Image of Mettler-Toledo Calibration substance ME 51143091, Saccharin
CAS Number:
Empirical Formula (Hill Notation):
 C7H5NO3S
Molecular Weight:
 183.18
 Beilstein/REAXYS Number:
 6888
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More