• USA Home
  • Search Results

  • mevastatin390517357388311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Mevastatin

2 Products
Synonym:   Compactin, ML-236B
Image of Mevastatin
CAS Number:
Empirical Formula (Hill Notation):
 C23H34O5
Molecular Weight:
 390.51
 Beilstein/REAXYS Number:
 1269441
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More