• USA Home
  • Search Results

  • mevinolinfromaspergillussp404547533075511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Mevinolin from Aspergillus sp.

1 Products
Synonym:   2-Methyl-1,2,3,7,8,8a-hexahydro-3,7-dimethyl-8-[2-(tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl)ethyl]-1-naphthalenyl ester butanoic acid, 6-α-Methylcompactin, Lovastatin, Monacolin K
Image of Mevinolin from <I>Aspergillus</I> sp.
CAS Number:
Empirical Formula (Hill Notation):
 C24H36O5
Molecular Weight:
 404.54
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
Show More