• USA Home
  • Search Results

  • mexiletinehydrochloride21572537001411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Mexiletine hydrochloride

3 Products
Synonym:   1-(2,6-Dimethylphenoxy)-2-propanamine hydrochloride, 1-(2,6-Xylyloxy)-2-aminopropane hydrochloride
Image of Mexiletine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C11H17NO · HCl
Molecular Weight:
 215.72
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Physical Form
  Special Grade
Show More