• USA Home
  • Search Results

  • mfluorodlphenylalanine1831845688211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

m-Fluoro-DL-phenylalanine

1 Products
Synonym:   3-Fluoro-DL-phenylalanine
Image of <I>m</I>-Fluoro-<SC>DL</SC>-phenylalanine
CAS Number:
Linear Formula:
 FC6H4CH2CH(NH2)COOH
Molecular Weight:
 183.18
 Beilstein/REAXYS Number:
 2939807
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
F5126 m-Fluoro-DL-phenylalanine,