• USA Home
  • Search Results

  • mfluorodltyrosine1991840390711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

m-Fluoro-DL-tyrosine

1 Products
Image of <I>m</I>-Fluoro-<SC>DL</SC>-tyrosine
CAS Number:
Linear Formula:
 FC6H3-4-(OH)CH2CH(NH2)CO2H
Molecular Weight:
 199.18
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
F4505 m-Fluoro-DL-tyrosine,