• USA Home
  • Search Results

  • miconazole416132291647811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Miconazole

3 Products
Synonym:   (±)-1-[2-(2,4-Dichlorobenzyloxy)-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole
CAS Number:
Empirical Formula (Hill Notation):
 C18H14Cl4N2O
Molecular Weight:
 416.13
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
Show More