• USA Home
  • Search Results

  • midodrinehydrochloride29074309217911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Midodrine hydrochloride

2 Products
Synonym:   (±)-1-(2,5-Dimethoxyphenyl)-2-glycinamidoethanol
Image of Midodrine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C12H18N2O4 · HCl
Molecular Weight:
 290.74
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Special Grade
Show More