• USA Home
  • Search Results

  • minocyclinehydrochloride493941361498711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Minocycline hydrochloride

5 Products
Image of Minocycline hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C23H27N3O7 · HCl
Molecular Weight:
 493.94
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
Show More