• USA Home
  • Search Results

  • miodobenzylguanidinehemisulfatesalt324138066395211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

m-Iodobenzylguanidine hemisulfate salt

1 Products
Synonym:   MIBG
Image of <I>m</I>-Iodobenzylguanidine hemisulfate salt
CAS Number:
Linear Formula:
 C8H10IN3 · 1/2H2SO4
Molecular Weight:
 324.13
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More