• USA Home
  • Search Results

  • modafinil273356869311811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Modafinil

2 Products
Synonym:   2-[(Diphenylmethyl)sulfinyl]­acetamide
Image of Modafinil
CAS Number:
Empirical Formula (Hill Notation):
 C15H15NO2S
Molecular Weight:
 273.35
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
Show More