• USA Home
  • Search Results

  • molecularsieves31234530808099111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Molecular sieves, 3 Å

4 Products
Synonym:   Molecular Sieve UOP Type 3Å
CAS Number:
Linear Formula:
 KnNa12-n[(AlO2)12(SiO2)12] · xH2O
 MDL number:
 eCl@ss:
 32110204
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More