• USA Home
  • Search Results

  • molybdenumhexacarbonyl264001393906511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Molybdenumhexacarbonyl

4 Products
Synonym:   Hexacarbonylmolybdenum(0)
Image of Molybdenumhexacarbonyl
CAS Number:
Linear Formula:
 Mo(CO)6
Molecular Weight:
 264.00
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More