• USA Home
  • Search Results

  • molybdenumivsulfide16007131733511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Molybdenum(IV) sulfide

3 Products
Synonym:   Molybdenum disulfide
Image of Molybdenum(IV) sulfide
CAS Number:
Empirical Formula (Hill Notation):
 MoS2
Molecular Weight:
 160.07
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Assay
  Particle Dimension
Show More